siRNA Details
| Gene Symbol: Asb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011984.1 | NP_001011984.1 | 6 | 851 | ||
| XM_006240464.2 | XP_006240526.1 | 4 | 1496 | |||
| GenBank mRNA | BC085882.1 | - | 6 | 851 | 7191983 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asb2| Gene Symbol: Asb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011984.1 | NP_001011984.1 | 6 | 851 | ||
| XM_006240464.2 | XP_006240526.1 | 4 | 1496 | |||
| GenBank mRNA | BC085882.1 | - | 6 | 851 | 7191983 | |