siRNA Details
| Gene Symbol: Gns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011989.1 | NP_001011989.1 | 3 | 320 | ||
| XM_006241396.2 | XP_006241458.1 | 3 | 416 | |||
| GenBank mRNA | BC087741.1 | - | 3 | 320 | 7314130 | |
| Gene Symbol: LOC100909505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750325.3 | XP_003750373.1 | 417 | |||

Gns
Genomic Map