siRNA Details
Gene Symbol: Tmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011997.1 | NP_001011997.1 | 3 | 309 | ||
GenBank mRNA | BC079290.1 | - | 3 | 309 | 7134366 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011997.1 | NP_001011997.1 | 3 | 309 | ||
GenBank mRNA | BC079290.1 | - | 3 | 309 | 7134366 |