siRNA Details
| Gene Symbol: FNBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024948.2 | NP_001020119.1 | 9 | 994 | ||
| NM_001164473.2 | NP_001157945.1 | 9 | 994 | |||
| NM_017737.4 | NP_060207.2 | 9 | 994 | |||
| XM_011541625.2 | XP_011539927.1 | 9 | 1058 | |||
| XM_017001533.1 | XP_016857022.1 | 10 | 1189 | |||
| GenBank mRNA | AK000282.1 | - | 3 | 246 | ||
| AK299760.1 | - | 4 | 380 | |||
| AK299943.1 | - | 3 | 290 | |||
| AY514449.1 | - | 9 | 843 | |||
| BC062477.1 | - | 4 | 445 | |||
| BC074891.2 | - | 3 | 224 | |||
| BC074892.2 | - | 3 | 224 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001114665, NM_153118, BC034510, BM964102.

FNBP1L
Genomic Map