siRNA Details
| Gene Symbol: Fbxo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011998.1 | NP_001011998.1 | 12 | 1524 | ||
| XM_006243419.2 | XP_006243481.1 | 12 | 1242 | |||
| GenBank mRNA | BC085831.1 | - | 12 | 1524 | 7134220 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo9| Gene Symbol: Fbxo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011998.1 | NP_001011998.1 | 12 | 1524 | ||
| XM_006243419.2 | XP_006243481.1 | 12 | 1242 | |||
| GenBank mRNA | BC085831.1 | - | 12 | 1524 | 7134220 | |