siRNA Details
| Gene Symbol: Morf4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011999.1 | NP_001011999.1 | 4 | 327 | ||
| XM_008766445.1 | XP_008764667.1 | 5 | 410 | |||
| GenBank mRNA | BC078910.1 | - | 4 | 327 | 7131534 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Morf4l1| Gene Symbol: Morf4l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011999.1 | NP_001011999.1 | 4 | 327 | ||
| XM_008766445.1 | XP_008764667.1 | 5 | 410 | |||
| GenBank mRNA | BC078910.1 | - | 4 | 327 | 7131534 | |