siRNA Details
| Gene Symbol: Lrrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012001.1 | NP_001012001.1 | 7, 8 | 994 | ||
| XM_006243926.2 | XP_006243988.1 | 7, 8 | 1201 | |||
| GenBank mRNA | BC085884.1 | - | 7, 8 | 994 | 7192145 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc2| Gene Symbol: Lrrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012001.1 | NP_001012001.1 | 7, 8 | 994 | ||
| XM_006243926.2 | XP_006243988.1 | 7, 8 | 1201 | |||
| GenBank mRNA | BC085884.1 | - | 7, 8 | 994 | 7192145 | |