siRNA Details
| Gene Symbol: Zap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012002.1 | NP_001012002.1 | 4 | 792 | ||
| XM_006244761.2 | XP_006244823.1 | 4 | 1931 | |||
| XM_008766998.1 | XP_008765220.1 | 4 | 1414 | |||
| GenBank mRNA | BC089855.1 | - | 4 | 792 | 7380367 | |

Zap70
Genomic Map