siRNA Details
| Gene Symbol: Rnf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012004.2 | NP_001012004.2 | 10 | 1369 | ||
| XM_006245172.2 | XP_006245234.1 | 10 | 1397 | |||
| XM_006245173.1 | XP_006245235.1 | 10 | 2250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf25| Gene Symbol: Rnf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012004.2 | NP_001012004.2 | 10 | 1369 | ||
| XM_006245172.2 | XP_006245234.1 | 10 | 1397 | |||
| XM_006245173.1 | XP_006245235.1 | 10 | 2250 | |||