siRNA Details
| Gene Symbol: Zfp18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012008.1 | NP_001012008.1 | 6 | 1500 | ||
| XM_006246695.2 | XP_006246757.1 | 7 | 1670 | |||
| XM_006246696.2 | XP_006246758.1 | 7 | 1667 | |||
| XM_006246697.2 | XP_006246759.1 | 6 | 1556 | |||
| GenBank mRNA | BC081874.1 | - | 6 | 1500 | 7107522 | |

Zfp18
Genomic Map