siRNA Details
| Gene Symbol: Snx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012012.2 | NP_001012012.2 | 8 | 1288 | ||
| NM_001199169.1 | NP_001186098.1 | 7 | 1228 | |||
| XM_006247195.2 | XP_006247257.1 | 9 | 1681 | |||
| XM_006247196.2 | XP_006247258.1 | 8 | 1458 | |||
| XM_006247198.2 | XP_006247260.1 | 7 | 1398 | |||
| XM_008768047.1 | XP_008766269.1 | 9 | 2032 | |||
| XM_008768048.1 | XP_008766270.1 | 9 | 2182 | |||
| GenBank mRNA | BC086543.1 | - | 8 | 1286 | 7132863 | |
| CO558022.1 | - | 2 | 627 | |||
| FQ212083.1 | - | 9 | 1342 | |||

Snx11
Genomic Map