siRNA Details
| Gene Symbol: Terf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012464.1 | NP_001012482.1 | 9 | 1073 | ||
| XM_006237729.2 | XP_006237791.1 | 8 | 1046 | |||
| GenBank mRNA | BC089888.1 | - | 9 | 1073 | 7374057 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Terf1| Gene Symbol: Terf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012464.1 | NP_001012482.1 | 9 | 1073 | ||
| XM_006237729.2 | XP_006237791.1 | 8 | 1046 | |||
| GenBank mRNA | BC089888.1 | - | 9 | 1073 | 7374057 | |