siRNA Details
| Gene Symbol: LINC01588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126496.1 | 1 | 836 | |||
| NR_126497.1 | 1 | 980 | ||||
| NR_126498.1 | 1 | 1837 | ||||
| NR_126500.1 | 1 | 980 | ||||
| GenBank mRNA | AK090420.1 | - | 1 | 836 | ||
| AK091232.1 | - | 1 | 1836 | |||
| BC128083.1 | - | 1 | 1559 | 40075904 | ||
| BC144351.1 | - | 1 | 979 | 9052871 | ||

LINC01588
Genomic Map