siRNA Details
| Gene Symbol: Clk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013041.1 | NP_001013059.1 | 6 | 737 | ||
| XM_006246234.2 | XP_006246296.1 | 7 | 947 | |||
| XM_006246236.1 | XP_006246298.1 | 10 | 1150 | |||
| XM_006246237.1 | XP_006246299.1 | 8 | 852 | |||
| XM_006246238.2 | XP_006246300.1 | 7 | 2367 | |||
| XM_008767670.1 | XP_008765892.1 | 8 | 1821 | |||
| XM_008767671.1 | XP_008765893.1 | 7 | 1743 | |||
| XM_008767672.1 | XP_008765894.1 | 7 | 786 | |||
| XM_008767673.1 | XP_008765895.1 | 6 | 2439 | |||
| XM_008767674.1 | XP_008765896.1 | 5 | 2226 | |||
| XM_008767675.1 | XP_008765897.1 | 8 | 1013 | |||
| XM_008767676.1 | XP_008765898.1 | 6 | 738 | |||
| GenBank mRNA | BC079006.1 | - | 6 | 737 | 7113075 | |
| FQ223275.1 | - | 9 | 2797 | |||

Clk4
Genomic Map