siRNA Details
| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013043.1 | NP_001013061.1 | 3, 4 | 523 | ||
| XM_006247885.2 | XP_006247947.1 | 3, 4 | 550 | |||
| GenBank mRNA | BC078937.1 | - | 3, 4 | 523 | 7108345 | |
| FQ225276.1 | - | 3, 4 | 516 | |||

Sectm1a
Genomic Map