siRNA Details
| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013044.1 | NP_001013062.1 | 13 | 1392 | ||
| GenBank mRNA | BC083853.1 | - | 13 | 1392 | 7124012 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cryzl1| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013044.1 | NP_001013062.1 | 13 | 1392 | ||
| GenBank mRNA | BC083853.1 | - | 13 | 1392 | 7124012 | |