siRNA Details
| Gene Symbol: Igfbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013048.1 | NP_001013066.1 | 5 | 859 | ||
| GenBank mRNA | BC086582.1 | - | 5 | 859 | 7307584 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igfbp7| Gene Symbol: Igfbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013048.1 | NP_001013066.1 | 5 | 859 | ||
| GenBank mRNA | BC086582.1 | - | 5 | 859 | 7307584 | |