siRNA Details
| Gene Symbol: Akr1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013057.1 | NP_001013075.1 | 10 | 1288 | ||
| XM_006254183.2 | XP_006254245.1 | 10 | 1315 | |||
| XM_006254184.2 | XP_006254246.1 | 10 | 1294 | |||
| XM_006254185.2 | XP_006254247.1 | 9 | 1237 | |||
| GenBank mRNA | BC078957.1 | - | 10 | 1288 | 7110806 | |

Akr1c2
Genomic Map