siRNA Details
| Gene Symbol: Rbm17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013058.1 | NP_001013076.1 | 8, 9 | 1005 | ||
| XM_006254205.2 | XP_006254267.1 | 8, 9 | 1112 | |||
| GenBank mRNA | BC079248.1 | - | 8, 9 | 1005 | 7132972 | |
| FQ230702.1 | - | 7, 8 | 865 | |||

Rbm17
Genomic Map