siRNA Details
| Gene Symbol: Rit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013060.1 | NP_001013078.1 | 5 | 1164 | ||
| GenBank mRNA | BC091382.1 | - | 5 | 1164 | 7320994 | |
| FQ213009.1 | - | 5 | 961 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rit2| Gene Symbol: Rit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013060.1 | NP_001013078.1 | 5 | 1164 | ||
| GenBank mRNA | BC091382.1 | - | 5 | 1164 | 7320994 | |
| FQ213009.1 | - | 5 | 961 | |||