siRNA Details
| Gene Symbol: KCTD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017634.3 | NP_060104.2 | 10 | 1089 | ||
| GenBank mRNA | AK000045.1 | - | 5 | 417 | ||
| BC021216.2 | - | 10 | 892 | 4302317 | ||
| BC068518.1 | - | 10 | 902 | 5265435 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCTD9| Gene Symbol: KCTD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017634.3 | NP_060104.2 | 10 | 1089 | ||
| GenBank mRNA | AK000045.1 | - | 5 | 417 | ||
| BC021216.2 | - | 10 | 892 | 4302317 | ||
| BC068518.1 | - | 10 | 902 | 5265435 | ||