siRNA Details
| Gene Symbol: Mrpl46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013068.1 | NP_001013086.1 | 3 | 530 | ||
| GenBank mRNA | BC086407.1 | - | 3 | 530 | 7306696 | |
| FQ212933.1 | - | 3 | 526 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl46| Gene Symbol: Mrpl46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013068.1 | NP_001013086.1 | 3 | 530 | ||
| GenBank mRNA | BC086407.1 | - | 3 | 530 | 7306696 | |
| FQ212933.1 | - | 3 | 526 | |||