siRNA Details
| Gene Symbol: Pknox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013074.1 | NP_001013092.1 | 10 | 1190 | ||
| XM_006256187.2 | XP_006256249.1 | 11 | 2225 | |||
| XM_006256188.2 | XP_006256250.1 | 12 | 2287 | |||
| XM_006256189.2 | XP_006256251.1 | 12 | 2279 | |||
| XM_006256190.2 | XP_006256252.1 | 11 | 2223 | |||
| XM_008772808.1 | XP_008771030.1 | 12 | 2102 | |||
| XM_008772809.1 | XP_008771031.1 | 11 | 2217 | |||
| XM_008772810.1 | XP_008771032.1 | 11 | 2105 | |||
| GenBank mRNA | BC091397.1 | - | 10 | 1190 | 7323315 | |

Pknox1
Genomic Map