siRNA Details
Gene Symbol: Plekha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013077.1 | NP_001013095.1 | 8 | 1287 | ||
GenBank mRNA | BC083759.1 | - | 8 | 1287 | 7192931 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plekha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013077.1 | NP_001013095.1 | 8 | 1287 | ||
GenBank mRNA | BC083759.1 | - | 8 | 1287 | 7192931 |