siRNA Details
| Gene Symbol: Pon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013082.1 | NP_001013100.1 | 9 | 1190 | ||
| XM_008762699.1 | XP_008760921.1 | 9 | 1267 | |||
| GenBank mRNA | BC079462.1 | - | 9 | 1190 | 7122087 | |
| FQ223399.1 | - | 1 | 30 | |||

Pon2
Genomic Map