siRNA Details
| Gene Symbol: Pon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013082.1 | NP_001013100.1 | 8, 9 | 966 | ||
| XM_008762699.1 | XP_008760921.1 | 8, 9 | 1043 | |||
| GenBank mRNA | BC079462.1 | - | 8, 9 | 966 | 7122087 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pon2| Gene Symbol: Pon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013082.1 | NP_001013100.1 | 8, 9 | 966 | ||
| XM_008762699.1 | XP_008760921.1 | 8, 9 | 1043 | |||
| GenBank mRNA | BC079462.1 | - | 8, 9 | 966 | 7122087 | |