siRNA Details
| Gene Symbol: Akr1b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013084.1 | NP_001013102.1 | 11 | 1134 | ||
| GenBank mRNA | BC079133.1 | - | 11 | 1134 | 7127413 | |
| Gene Symbol: LOC100910708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762861.1 | XP_008761083.1 | 10 | 1282 | ||

Akr1b10
Genomic Map