siRNA Details
| Gene Symbol: Elmod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013087.1 | NP_001013105.1 | 11 | 1141 | ||
| XM_006236667.2 | XP_006236729.1 | 11 | 1141 | |||
| XM_006236669.1 | XP_006236731.1 | 10 | 1093 | |||
| XM_008762978.1 | XP_008761200.1 | 10 | 1103 | |||
| GenBank mRNA | BC083623.1 | - | 11 | 1141 | 7134106 | |

Elmod3
Genomic Map