siRNA Details
| Gene Symbol: Slc38a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013099.1 | NP_001013117.1 | 2 | 288 | ||
| XM_006240209.1 | XP_006240271.1 | 2 | 278 | |||
| XM_006240210.2 | XP_006240272.1 | 2 | 311 | |||
| GenBank mRNA | AY266018.1 | - | 2 | 288 | ||

Slc38a6
Genomic Map