siRNA Details
| Gene Symbol: Abcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013100.3 | NP_001013118.2 | 14, 15 | 1515 | ||
| XM_006240330.2 | XP_006240392.1 | 14, 15 | 1602 | |||
| XM_006240331.2 | XP_006240393.1 | 15 | 1750 | |||
| GenBank mRNA | BC089214.1 | - | 7, 8 | 1066 | ||

Abcd4
Genomic Map