siRNA Details
| Gene Symbol: Abcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013100.3 | NP_001013118.2 | 19 | 1882 | ||
| XM_006240330.2 | XP_006240392.1 | 19 | 1969 | |||
| GenBank mRNA | BC089214.1 | - | 12 | 1433 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcd4| Gene Symbol: Abcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013100.3 | NP_001013118.2 | 19 | 1882 | ||
| XM_006240330.2 | XP_006240392.1 | 19 | 1969 | |||
| GenBank mRNA | BC089214.1 | - | 12 | 1433 | ||