siRNA Details
| Gene Symbol: Fkbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC084709.1 | - | 3 | 436 | 7301886 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fkbp11| Gene Symbol: Fkbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC084709.1 | - | 3 | 436 | 7301886 | |