siRNA Details
| Gene Symbol: Gna13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013119.1 | NP_001013137.1 | 2 | 459 | ||
| GenBank mRNA | AY553631.1 | - | 2 | 459 | ||
| AY553632.1 | - | 2 | 459 | |||
| DQ120481.1 | - | 2 | 459 | |||
| DQ120482.1 | - | 2 | 459 | |||

Gna13
Genomic Map