siRNA Details
| Gene Symbol: Timd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013855.1 | NP_001013877.1 | 4, 5 | 726 | ||
| XM_006246168.1 | XP_006246230.1 | 5, 6 | 1011 | |||
| XM_008767667.1 | XP_008765889.1 | 5, 6 | 774 | |||
| XM_008767668.1 | XP_008765890.1 | 6, 7 | 741 | |||
| XM_008767669.1 | XP_008765891.1 | 5, 6 | 1266 | |||
| GenBank mRNA | BC089832.1 | - | 4, 5 | 726 | 7324518 | |

Timd2
Genomic Map