siRNA Details
| Gene Symbol: Cnot6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013856.1 | NP_001013878.1 | 4 | 415 | ||
| GenBank mRNA | BC079308.1 | - | 4 | 415 | 7134677 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnot6| Gene Symbol: Cnot6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013856.1 | NP_001013878.1 | 4 | 415 | ||
| GenBank mRNA | BC079308.1 | - | 4 | 415 | 7134677 | |