siRNA Details
| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013858.1 | NP_001013880.1 | 4 | 628 | ||
| XM_008767957.1 | XP_008766179.1 | 4 | 788 | |||
| GenBank mRNA | BC085876.1 | - | 4 | 628 | 7191659 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tlcd1| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013858.1 | NP_001013880.1 | 4 | 628 | ||
| XM_008767957.1 | XP_008766179.1 | 4 | 788 | |||
| GenBank mRNA | BC085876.1 | - | 4 | 628 | 7191659 | |