siRNA Details
| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013866.1 | NP_001013888.1 | 6 | 571 | ||
| GenBank mRNA | BC089826.1 | - | 6 | 571 | 7321402 | |
| FQ214930.1 | - | 6 | 577 | |||
| FQ219915.1 | - | 6 | 572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmss1| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013866.1 | NP_001013888.1 | 6 | 571 | ||
| GenBank mRNA | BC089826.1 | - | 6 | 571 | 7321402 | |
| FQ214930.1 | - | 6 | 577 | |||
| FQ219915.1 | - | 6 | 572 | |||