siRNA Details
Gene Symbol: LOC289035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013872.1 | NP_001013894.2 | 5 | 1331 | ||
GenBank mRNA | BC079259.1 | - | 5 | 1331 | 7133407 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC289035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013872.1 | NP_001013894.2 | 5 | 1331 | ||
GenBank mRNA | BC079259.1 | - | 5 | 1331 | 7133407 |