siRNA Details
| Gene Symbol: Clhc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013877.1 | NP_001013899.1 | 9 | 1296 | ||
| XM_008770455.1 | XP_008768677.1 | 9 | 1417 | |||
| GenBank mRNA | BC083604.1 | - | 9 | 1296 | 7133284 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clhc1| Gene Symbol: Clhc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013877.1 | NP_001013899.1 | 9 | 1296 | ||
| XM_008770455.1 | XP_008768677.1 | 9 | 1417 | |||
| GenBank mRNA | BC083604.1 | - | 9 | 1296 | 7133284 | |