siRNA Details
| Gene Symbol: RGD1311847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013879.1 | NP_001013901.1 | 8 | 2212 | ||
| GenBank mRNA | BC079430.1 | - | 8 | 2212 | 7114707 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311847| Gene Symbol: RGD1311847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013879.1 | NP_001013901.1 | 8 | 2212 | ||
| GenBank mRNA | BC079430.1 | - | 8 | 2212 | 7114707 | |