siRNA Details
| Gene Symbol: Gatad2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013881.2 | NP_001013903.2 | 8 | 1369 | ||
| XM_006252879.2 | XP_006252941.1 | 7 | 1049 | |||
| XM_006252880.2 | XP_006252942.1 | 7 | 1046 | |||
| XM_006252881.2 | XP_006252943.1 | 7 | 1040 | |||
| XM_006252882.2 | XP_006252944.1 | 7 | 1037 | |||
| XM_006252883.2 | XP_006252945.1 | 7 | 1049 | |||
| XM_006252884.2 | XP_006252946.1 | 8 | 1328 | |||
| XM_006252887.2 | XP_006252949.1 | 7 | 861 | |||
| XM_006252889.2 | XP_006252951.1 | 7 | 1047 | |||
| XM_008771089.1 | XP_008769311.1 | 9 | 1418 | |||
| XM_008771090.1 | XP_008769312.1 | 8 | 1048 | |||
| GenBank mRNA | BC089902.1 | - | 5 | 459 | 7383529 | |

Gatad2a
Genomic Map