siRNA Details
| Gene Symbol: Dctd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013882.2 | NP_001013904.2 | 4 | 474 | ||
| GenBank mRNA | BC087138.1 | - | 4 | 429 | 7125992 | |
| DY317307.1 | - | 4 | 442 | |||
| EV776012.1 | - | 4 | 499 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dctd| Gene Symbol: Dctd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013882.2 | NP_001013904.2 | 4 | 474 | ||
| GenBank mRNA | BC087138.1 | - | 4 | 429 | 7125992 | |
| DY317307.1 | - | 4 | 442 | |||
| EV776012.1 | - | 4 | 499 | |||