siRNA Details
| Gene Symbol: Dctd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013882.2 | NP_001013904.2 | 5 | 630 | ||
| NM_001161512.1 | NP_001154984.1 | 4 | 749 | |||
| GenBank mRNA | BC087138.1 | - | 5 | 585 | 7125992 | |
| BP504069.1 | - | 751 | ||||
| DY317307.1 | - | 5 | 598 | |||
| EV776012.1 | - | 5 | 655 | |||

Dctd
Genomic Map