siRNA Details
| Gene Symbol: Ttc29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013890.1 | NP_001013912.1 | 7 | 789 | ||
| GenBank mRNA | BC078968.1 | - | 7 | 789 | 7111580 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc29| Gene Symbol: Ttc29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013890.1 | NP_001013912.1 | 7 | 789 | ||
| GenBank mRNA | BC078968.1 | - | 7 | 789 | 7111580 | |