siRNA Details
Gene Symbol: Txnl4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013891.1 | NP_001013913.1 | 2 | 342 | ||
XM_006255579.2 | XP_006255641.1 | 3 | 452 | |||
GenBank mRNA | BC089962.1 | - | 2 | 342 | 7314335 |
