siRNA Details
| Gene Symbol: l7Rn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013897.1 | NP_001013919.1 | 5 | 769 | ||
| XM_006229662.2 | XP_006229724.1 | 4 | 736 | |||
| XM_008759638.1 | XP_008757860.1 | 4 | 567 | |||
| XM_008759639.1 | XP_008757861.1 | 3 | 496 | |||
| GenBank mRNA | BC088340.1 | - | 5 | 769 | 7367791 | |
| FQ212130.1 | - | 4 | 680 | |||
| FQ213824.1 | - | 4 | 681 | |||
| FQ214677.1 | - | 4 | 681 | |||
| FQ215506.1 | - | 4 | 672 | |||
| FQ216197.1 | - | 4 | 681 | |||
| FQ216668.1 | - | 4 | 681 | |||
| FQ216759.1 | - | 4 | 681 | |||
| FQ218863.1 | - | 4 | 678 | |||
| FQ220044.1 | - | 4 | 681 | |||
| FQ220310.1 | - | 4 | 732 | |||
| FQ220575.1 | - | 4 | 681 | |||
| FQ224272.1 | - | 4 | 683 | |||
| FQ224945.1 | - | 4 | 681 | |||
| FQ229506.1 | - | 4 | 677 | |||

l7Rn6
Genomic Map