siRNA Details
Gene Symbol: RGD1311703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089924.1 | - | 5 | 1081 | 7307309 | |
FQ225875.1 | - | 5 | 1118 | |||
FQ227306.1 | - | 5 | 1113 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1311703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089924.1 | - | 5 | 1081 | 7307309 | |
FQ225875.1 | - | 5 | 1118 | |||
FQ227306.1 | - | 5 | 1113 |