siRNA Details
Gene Symbol: RGD1311703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230086.2 | XP_006230148.1 | 5 | 703 | ||
GenBank mRNA | BC089924.1 | - | 5 | 668 | 7307309 | |
FQ227306.1 | - | 5 | 701 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1311703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230086.2 | XP_006230148.1 | 5 | 703 | ||
GenBank mRNA | BC089924.1 | - | 5 | 668 | 7307309 | |
FQ227306.1 | - | 5 | 701 |