siRNA Details
| Gene Symbol: ZNF280C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017666.4 | NP_060136.1 | 4 | 428 | ||
| XM_006724765.3 | XP_006724828.1 | 4 | 379 | |||
| GenBank mRNA | AK000102.1 | - | 4 | 351 | ||
| AK290373.1 | - | 4 | 379 | |||
| AL133204.1 | - | 4 | 363 | |||
| AL834333.1 | - | 4 | 359 | |||
| BC051728.1 | - | 4 | 379 | 4828435 | ||
| DR003059.1 | - | 4 | 427 | |||

ZNF280C
Genomic Map