siRNA Details
| Gene Symbol: Pagr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013901.1 | NP_001013923.1 | 2 | 1285 | ||
| GenBank mRNA | BC088203.1 | - | 2 | 1285 | 7379842 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pagr1| Gene Symbol: Pagr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013901.1 | NP_001013923.1 | 2 | 1285 | ||
| GenBank mRNA | BC088203.1 | - | 2 | 1285 | 7379842 | |